Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TXNL1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299643PCP
Description
TXNL1 Polyclonal antibody specifically detects TXNL1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
TXNL1 | |
Polyclonal | |
Western Blot, Immunohistochemistry-Paraffin | |
32 kDa thioredoxin-related protein, thioredoxin-like 1, thioredoxin-like, 32kDa, thioredoxin-related 32 kDa protein, thioredoxin-related protein 1, TRP32thioredoxin-like protein 1, Txl, TXL-1, TXNL | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human TXNL1. | |
0.1 mL | |
Apoptosis | |
9352 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction