Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TXNL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531294
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531294 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531294 Supplier Invitrogen™ Supplier No. PA531294
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Raji. Predicted reactivity: Mouse (92%), Rat (92%), Xenopus laevis (80%), Pig (90%), Chicken (83%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the glutaredoxin family. Glutaredoxins are oxidoreductase enzymes that reduce a variety of substrates using glutathione as a cofactor. The encoded protein binds to and modulates the function of protein kinase C theta. The encoded protein may also inhibit apoptosis and play a role in cellular growth, and the expression of this gene may be a marker for cancer. Pseudogenes of this gene are located on the short arm of chromosomes 6 and 9. Alternatively spliced transcript variants have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNL2
Applications Western Blot
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene GLRX3
Gene Accession No. O76003
Gene Alias bA500G10.4; FLJ11864; Glrx3; GLRX4; glutaredoxin 3; glutaredoxin 4; glutaredoxin-3; GRX3; GRX4; HUSSY-22; Picot; PKC interacting cousin of thioredoxin; PKC-interacting cousin of thioredoxin; PKCq-interacting protein; PKC-theta-interacting protein; testicular tissue protein Li 75; thioredoxin-like 2; thioredoxin-like protein 2; TXNL2; TXNL3
Gene Symbols GLRX3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TXNL2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10539
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.