Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TXNL4A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576132
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Essential role in pre-mRNA splicing as component of the U5 snRNP and U4/U6-U5 tri-snRNP complexes that are involved in spliceosome assembly.
Specifications
TXNL4A | |
Polyclonal | |
Unconjugated | |
TXNL4A | |
BMKS; D18Wsu98e; DIB1; Dim1; DIM1 protein homolog; ENSMUSG00000057130; SNRNP15; spliceosomal U5 snRNP-specific 15 kDa protein; thioredoxin like 4A; thioredoxin-like 4; thioredoxin-like 4A; thioredoxin-like protein 4A; Thioredoxin-like U5 snRNP protein U5-15kD; TXNL4; Txnl4a; U5-15kD; U5-15kDa | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10907, 27366 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P83876, P83877 | |
TXNL4A | |
Recombinant fusion protein of human TXNL4A. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction