Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TXNL4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145128
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145128 Supplier Invitrogen™ Supplier No. PA5145128
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DITEVPDFNK MYELYDPCTV MFFFRNKHIM IDLGTGNNNK INWAMEDKQE MVDIIETVYR G Antigen sequence identity: Mouse 100%; Rat 100%.

Essential role in pre-mRNA splicing as component of the U5 snRNP and U4/U6-U5 tri-snRNP complexes that are involved in spliceosome assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNL4A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TXNL4A
Gene Accession No. P83876
Gene Alias BMKS; D18Wsu98e; DIB1; Dim1; DIM1 protein homolog; ENSMUSG00000057130; SNRNP15; spliceosomal U5 snRNP-specific 15 kDa protein; thioredoxin like 4A; thioredoxin-like 4; thioredoxin-like 4A; thioredoxin-like protein 4A; Thioredoxin-like U5 snRNP protein U5-15kD; TXNL4; Txnl4a; U5-15kD; U5-15kDa
Gene Symbols TXNL4A
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 62-122. Recombinant protein control fragment (Product #RP-110141).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10907
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.