Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNL4A Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735726 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5726 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5726 Supplier Proteintech Group Inc Supplier No. 276461AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Essential role in pre-mRNA splicing as component of the U5 snRNP and U4/U6-U5 tri-snRNP complexes that are involved in spliceosome assembly.

Specifications

Antigen TXNL4A
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TXNL4A
Gene Accession No. P83876
Gene Alias DIB1, DIM1, DIM1 protein homolog, HsT161, thioredoxin like 4A, Thioredoxin like protein 4A, TXNL4, TXNL4A, U5 15kD
Gene Symbols TXNL4A
Host Species Rabbit
Immunogen TXNL4A Fusion Protein Ag26473
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10907
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.