Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TXNL4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596584
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596584 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596584 Supplier Invitrogen™ Supplier No. PA596584
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSFLLPKLTS KKEVDQAIKS TAEKVLVLRF GRDEDPVCLQ LDDILSKTSS DLSKMAAIYL VDVDQTAVYT QYFDISYIPS TVFFFNGQHM KVDYGSPDHT KFVGSFKTKQ DFIDLIEVIY RGAMRGKLIV QSPIDPKNIP KYDLLYQDI; Positive Samples: Mouse liver; Cellular Location: Nucleus.

Dim2, also known as TXNL4B (thioredoxin-like 4B), or DLP, is a 149 amino acid nuclear protein that exists as a homodimer and plays an essential role in pre-mRNA splicing. Evolutionarily related and sharing 38% sequence identity with Dim1, Dim2 is required for S/G(2) transition during the cell cycle and is able to bind the PRP6 (U5-102K) subunit of the spliceosome. The gene encoding Dim2 maps to human chromosome 16, which encodes over 900 genes and comprises nearly 3% of the human genome. The rare disorder Rubinstein-Taybi syndrome is associated with chromosome 16, as is Crohn's disease, which is a gastrointestinal inflammatory condition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNL4B
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TXNL4B
Gene Accession No. Q8BUH1, Q9NX01
Gene Alias AI595343; D530025J19Rik; Dim1-like protein; DIM2; DLP; RGD1305127; thioredoxin like 4B; thioredoxin-like 4B; thioredoxin-like protein 4B; Txnl4b
Gene Symbols TXNL4B
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234723, 54957
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.