Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TXNL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547535
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547535 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547535 Supplier Invitrogen™ Supplier No. PA547535
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody detects endogenous mouse and rat Thioredoxin-like 5/TRP14 in Western blots. Reconstitute at 0.2 mg/mL in sterile PBS.

TXNL5 disulfide reductase. May participate in various redox reactions through the reversible oxidation of its active center dithiol to a disulfide and catalyze dithiol-disulfide exchange reactions. Modulates TNF-alpha signaling and NF-kappa-B activation. Has peroxidase activity and may contribute to the elimination of cellular hydrogen peroxide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNL5
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene TXNDC17
Gene Accession No. Q9CQM5
Gene Alias 14 kDa thioredoxin-related protein; 4831443O22Rik; D11Ertd672e; MGC14353; protein 42-9-9; putative 42-9-9 protein; testicular tissue protein Li 214; thioredoxin (Trx)-related protein, 14 kDa; thioredoxin domain containing 17; thioredoxin domain-containing protein 17; thioredoxin-like 5; thioredoxin-like protein 5; TRP14; Txndc17; Txnl5
Gene Symbols TXNDC17
Host Species Goat
Immunogen E. coli-derived recombinant mouse Thioredoxin-like 5/TRP14 Met1-Asp123.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287474, 52700
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.