Learn More
TXNRD1 Mouse anti-Human, Mouse, Rat, Clone: 1B10C4, ProteinTech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 677281IG150UL

Description
Aliquoting is unnecessary for -20°C storage.
TXNRD1 Monoclonal antibody specifically detects TXNRD1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
TXNRD1 | |
Monoclonal | |
294 μg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
O89049, Q16881, Q9JMH6 | |
TXNRD1 | |
TXNRD1 fusion protein Ag30071 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1B10C4 | |
Unconjugated | |
TXNRD1 | |
gene associated with retinoic and IFN-induced mortality 12 protein; gene associated with retinoic and interferon-induced mortality 12 protein; GRIM12; GRIM-12; KDRF; KM-102-derived reductase-like factor; MGC9145; NADPH-dependent thioredoxin reductase; oxidoreductase; selenoprotein oxidoreductase; testis tissue sperm-binding protein Li 46a; thioredoxin reductase 1; thioredoxin reductase 1, cytoplasmic; thioredoxin reductase GRIM-12; thioredoxin reductase TR1; TR; TR alpha; TR1; Trxr1; TXNR; Txnrd1 | |
Mouse | |
Protein G | |
RUO | |
50493, 58819, 7296 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.