Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNRD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565167
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565167 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565167 Supplier Thermo Scientific Supplier No. PA565167
Add to Cart
Add to Cart

TXNRD3 Polyclonal Antibody

TXNRD3 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNRD3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TXNRD3
Gene Accession No. Q86VQ6
Gene Alias TGR; Thioredoxin and glutathione reductase; thioredoxin glutathione reductase; thioredoxin reductase 2; thioredoxin reductase 3; Thioredoxin reductase TR2; TR2; TRXR3; TXNRD3
Gene Symbols TXNRD3
Host Species Rabbit
Immunogen Recombinant Human TXNRD3. Recombinant protein control fragment (Product #RP-105917).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114112
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.