Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TY 52156
GREENER_CHOICE

Catalog No. 532810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-281-0 10 mg
53-285-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-281-0 Supplier Tocris Bioscience™ Supplier No. 5328/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

S1P3 receptor antagonist

TY 52156 is an S1P3 receptor antagonist (Ki = 110 nM); inhibits S1P-induced Ca2+ release in HUVEC cells. Suppresses FTY-720-induced S1P3 receptor-mediated bradycardia in vivo. Also inhibits S1P-induced breast cancer stem cell expansion in vitro. Orally bioavailable.

Specifications

Quantity 10 mg
Product Type TY 52156
Purity >98%
Molecular Weight (g/mol) 364.27

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.