Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ty1-tag Rabbit, Polyclonal, GenScript™

Catalog No. 50273135
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-273-135 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-273-135 Supplier Genscript Corporation Supplier No. A0100440
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL. Well-characterized antibodies for epitope tags are common in the study of protein expression in various systems.GenScript Rabbit Anti- Ty1-tag Polyclonal Antibody is developed in rabbit using a KLH-coupled synthetic peptide.

Ty1-tag Polyclonal antibody specifically detects Ty1-tag in Tag samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen Ty1-tag
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene Alias Ty1 Epitope Tag
Host Species Rabbit
Immunogen Ty1-tag epitope peptide conjugated to KLH
Purification Method Affinity chromatography
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.