Learn More
DuPont™ Tychem™ 10000 Coverall with Respirator Hood, Elastic Wrists, Sock Boots
Catalog No. p-7139611 Shop All DuPont Personal Protection Products
Click to view available options
Size:
Small
5X-Large
6X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Tychem™ 10000 offers a high level of protection against toxic, corrosive gases and liquid chemicals
Tychem™ 10000 has excellent chemical barrier properties and high strength for enhanced durability. Developed for protection against toxic, corrosive gases and liquid chemicals, this high-performance chemical barrier fabric is suited for industrial, HAZMAT, and domestic preparedness applications. Tychem™ 10000 garments are available to meet NFPA 1991 and NFPA 1994 standards.
  • Provides at least 30 minutes of protection against over 320 chemical challenges
  • Chosen by HAZMAT responders worldwide
  • Available in high-visibility lime yellow
  • Garments include fully encapsulated, vapor protective Level A suit and liquid-splash protective Level B suits as well as coveralls.
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Material Tychem
Size 5X-Large
Ankle Style Boot Overflaps
Cuff Style Elastic
Disposable Single-use
Gender Unisex
Latex Free Yes
Neck Style Hooded
No. of Pockets None
Quantity 2/Pk.
Resistance Liquid
Seam Type Taped
Sterility Non-sterile
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.