Learn More
DuPont™ Tychem™ 10000 Encapsulated Level A Suit with Viton™
Catalog No. 18888852 Shop All DuPont Personal Protection Products
Click to view available options
Size:
Medium
Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
Catalog No. 18-888-852
Supplier: DuPont™ TK554TLYLG00015C
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Encapsulated Level A garment offers a high level of protection against toxic, corrosive gases and liquid chemicals
  • 10000 Encapsulated Level A Suit with Viton™ design provides protection from liquid splash exposure for both the wearer and respiratory equipment
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Also provides coverage from overhead liquid splash (does not provide vapor protection)
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Standard 40 mil PVC faceshield
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Liquid resistant, extra long zipper to increase garment opening and aid donning and doffing
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
TRUSTED_SUSTAINABILITY
Specifications
Color Lime Yellow
Material Tychem
Size Large
Ankle Style Boot Overflaps
Closure Type Zipper
Seam Type Taped
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.