Learn More
DuPont™ Tychem™ 10000 Encapsulated Level A Suit with Viton Showa Best™ 890 Outer Gloves
Catalog No. 18888865 Shop All DuPont Personal Protection Products
This item is not returnable. View return policy
This item is not returnable. View return policy
Tychem™ 10000 offers a high level of protection against toxic, corrosive gases and liquid chemicals
Tychem™ 10000 has excellent chemical barrier properties and high strength for enhanced durability. Developed for protection against toxic, corrosive gases and liquid chemicals, this high-performance chemical barrier fabric is suited for industrial, HAZMAT, and domestic preparedness applications. Tychem™ 10000 garments are available to meet NFPA 1991 and NFPA 1994 standards.
  • Provides at least 30 minutes of protection against over 320 chemical challenges
  • Chosen by HAZMAT responders worldwide
  • Available in high-visibility lime yellow
  • Garments include fully encapsulated, vapor protective Level A suit and liquid-splash protective Level B suits as well as coveralls.
TRUSTED_SUSTAINABILITY
Specifications
Material Tychem
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.