Learn More
DuPont™ Tychem™ 2000 Series 125 Coveralls
Catalog No. 19067017
Click to view available options
Seam Type:
Bound
Serged
Taped
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Flexible and durable light liquid splash protection
Tychem™ QC fabric provides at least 30 minutes of protection against 42 chemical challenges. It is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing.
  • Lightweight and durable fabric
  • Laydown collar
  • Elastic opening for tighter fit at wrist and ankle
  • Serged seams
Compliance
Meets ASTM™ D3776, ASTM D1777, ASTM 1117, ASTM D3786, ASTM D5034 standards.
TRUSTED_SUSTAINABILITY
Specifications
Size 4X-Large
Material Tychem
Color Yellow
Seam Type Serged
Width (English) Chest 32 in.
Height (English) 6 ft. 4 in. to 6 ft. 7 in.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.