Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tychem™ 2000 SFR Apron
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Tychem™ 2000 SFR represents a new generation of secondary flame-resistant chemical garment technology
- Unlike traditional secondary flame resistant chemical garments that have been available for years, new Tychem™ 2000 SFR garments were specially designed to meet dual hazard needs of a protective chemical suit with secondary flame resistance
- This unique combination of performance permits Tychem™ 2000 SFR garments to be worn over primary flame resistant (FR) garments like Nomex™ when chemical splash and flash fire hazards exist
- The fabric used in Tychem™ 2000 SFR garments utilize a unique technology so that it does not char like traditional secondary flame resistant technologies - it is designed to shrink away from flame without burning
- Tychem™ 2000 SFR garments also provide an effective barrier against a range of inorganic acids, bases, industrial cleaning chemicals, and particles
- They're an ideal option for keeping workers safe from chemicals, while not compromising the flame protection of fire-resistance clothing worn beneath them
- Applications include: refineries, petrochemical plants, laboratories, hazardous maintenance operations

Specifications
Specifications
Size | X-Large |
Color | Green |
Disposable | Single-use |
Closure Type | Hook and Loop, Waist Ties |
Sterility | Non-sterile |
Cuff Style | Elastic |
Latex Free | Yes |
Thickness | 7.5 mil |
Length (English) | 45 in. |
Length (English) Cuff | 32.25 in. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction