Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tychem™ 4000 Coverall with Collar, Elastic Wrists, Attached Socks
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Description
DuPont™ Tyvek™ 400 provides light-weight inherent barrier protection against hazardous dry particles and aerosols, and nonhazardous light liquid splash.
- Excellent abrasion resistance due its unique, nonwoven material
- Can be used in a variety of applications including general maintenance/operations, asbestos and lead abatement, mold remediation, and environmental cleanups

Specifications
Specifications
Size | 5X-Large |
Gender | Unisex |
Color | White |
Disposable | Single-use |
Ankle Style | Attached Socks |
Cuff Style | Elastic |
Neck Style | Collared |
Closure Type | Storm Flap with Adhesive Closure |
Seam Type | Bound |
Sterility | Non-sterile |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction