Learn More
DuPont™ Tychem™ 9000 Collared Coverall
Click to view available options
Size:
Small
Medium
3X-Large
4X-Large
5X-Large
Description
DuPont™ Tychem™ 9000 is strong and durable for consistent and lasting protection.
- NFPA 1994 Class 2 and NFPA 1992 certified
- Provides at least 30 minutes of protection against over 280 chemical challenges
- Used by many in the chemical handling and petrochemical manufacturing
Specifications
Specifications
Size | 5X-Large |
Color | Yellow |
Ankle Style | Elastic |
Cuff Style | Elastic |
Closure Type | Storm Flap with Adhesive Closure |
Seam Type | Taped |