Learn More
DuPont™ Tychem™ ThermoPro™ Sleeveless Gray Aprons
Catalog No. p-7122030
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Designed to provide protection for industrial workers and Hazmat responders. DuPont™ Tychem™ ThermoPro™ Sleeveless Aprons provide permeation protection against a broad range of toxic industrial chemicals and chemical warfare agents
  • Tychem™ ThermoPro for triple hazard protection
  • Sleeveless apron design provides frontal body protection
  • For flash fire and electric arc hazards, partial body protective Tychem ThermoPro garments must be used in conjunction with primary flame resistant clothing that is rated for the fire/arc hazard.
  • Sewn with Nomex™ thread
  • Designed with Nomex waist strap and flame resistant (FR) nylon buckle
  • Designed with end-user input for laboratory applications
  • Ideal for use on top of an existing flame resistant (FR) garment where extra chemical protection is required
  • Gray color for discretionary needs
  • Made in the USA
Order Info
Ansell™ Vinyl Closed-Back Aprons, Waist ties
Compliance
North American Free Trade Agreement (NAFTA) and Trade Agreement Act (TAA).
TRUSTED_SUSTAINABILITY
Specifications
Size Small
Color Gray
For Use With (Application) HazMat Teams in Fire Departments, Industrial Fire Brigades, Clandestine Lab Investigation, Industrial Chemical Processing Plants, Laboratories
Seam Type Taped
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.