Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TYK2 Monoclonal Antibody (8G8E9, 8G9H5,8G8B3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515303
Description
MA5-15303 targets Tyk2 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15303 immunogen is purified recombinant fragment of Tyk2 expressed in E. Coli.
Tyrosine Kinase 2 (TYK2) a member of the JAK kinase family, plays both structural and catalytic roles in type I interferon (IFN) signaling. As such, it may play a role in anti-viral immunity. A mutation in this gene has been associated with hyperimmunoglobulin E syndrome (HIES) - a primary immunodeficiency characterized by elevated serum immunoglobulin E.
Specifications
TYK2 | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.03% sodium azide | |
P29597 | |
TYK2 | |
Purified recombinant fragment of Tyk2 expressed in E. Coli. | |
RUO | |
7297 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
8G8E9, 8G9H5,8G8B3, 8G9H5, 8G8B3 | |
Unconjugated | |
TYK2 | |
4833430L21Rik; AW540162; BRE1; BRE1A; C79397; hBRE1; IMD35; JTK1; mKIAA4116; Non-receptor tyrosine-protein kinase TYK2; Tyk2; tyrosine kinase 2; tyrosine kinase TYK2 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction