Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TYK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121841
This item is not returnable.
View return policy
Description
TYK2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Tyrosine Kinase 2 (TYK2) a member of the JAK kinase family, plays both structural and catalytic roles in type I interferon (IFN) signaling. As such, it may play a role in anti-viral immunity. A mutation in this gene has been associated with hyperimmunoglobulin E syndrome (HIES) - a primary immunodeficiency characterized by elevated serum immunoglobulin E.
Specifications
TYK2 | |
Polyclonal | |
Unconjugated | |
TYK2 | |
4833430L21Rik; AW540162; BRE1; BRE1A; C79397; hBRE1; IMD35; JTK1; mKIAA4116; Non-receptor tyrosine-protein kinase TYK2; Tyk2; tyrosine kinase 2; tyrosine kinase TYK2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54721 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9R117 | |
TYK2 | |
Recombinant protein Tyrosine Kinase 2. The antigen corresponds to amino acid range 667-914 of the target protein. | |
20 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction