Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYKi Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282045B
This item is not returnable.
View return policy
Description
TYKi Polyclonal antibody specifically detects TYKi in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
TYKi | |
Polyclonal | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
Biotin | |
cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial, cytidylate kinase 2, EC 2.7.4.14, mitochondrial, mitochondrial UMP-CMP kinase, thymidine monophosphate kinase 2, thymidylate kinase family LPS-inducible member, TMPK2, TYKi, UMP-CMPK2 | |
TYKi antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human TYKi. The immunogen is located within amino acids 370 - 420 of TYKi. Amino Acid Squence: PEERLQRLQGRGMEKTREE | |
0.1 mL | |
Protein Kinase | |
129607 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction