Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYMP Monoclonal Antibody (OTI7B4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA801109

Description
Platelet - derived endothelial growth factor (PD-ECGF), same as thymidine phosphorylase (TP) or gliostatin. In the presence of inorganic orthophosphate, it catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their correspondingbases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. It is also invoved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of maligcies, into active cytotoxic metabolites.Specifications
TYMP | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P19971 | |
TYMP | |
Human recombit protein fragment corresponding to amino acids 311-482 of human TYMP produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7B4 | |
Unconjugated | |
TYMP | |
ECGF, ECGF1, MEDPS1, MNGIE, MTDPS1, PDECGF, TP, hPD-ECGF | |
Mouse | |
Affinity Chromatography | |
RUO | |
1890 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction