Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYMP Mouse anti-Human, Clone: SPM322, Abnova™

Catalog No. 89034591 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-591 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-591 Supplier Abnova Corporation Supplier No. MAB14508
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant human TYMP.

  • High specificity
  • High reactivity

Specifications

Antigen TYMP
Applications Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone SPM322
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human TYMP.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Immunoprecipitation (0.5-1 ug/500 ug protein lysate) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. P19971
Gene Alias ECGF1/MNGIE/PDECGF/TP/hPD-ECGF
Gene Symbols TYMP
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human TYMP.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1890
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.