Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYMS Mouse anti-Human, HRP, Clone: 1F5B7, Proteintech
SDP

Catalog No. 501737244 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7244 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7244 Supplier Proteintech Group Inc Supplier No. HRP66725100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thymidylate Synthase (TS) (EC 2.1.1.45) converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. TS is a critical target for the fluoropyrimidines, an important group of antineoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates TS by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. Expression of TS protein is associated with response to 5-fluorouracil (5-FU) in human colorectal, gastric, head and neck, and breast carcinomas.

Specifications

Antigen TYMS
Applications Western Blot
Classification Monoclonal
Clone 1F5B7
Concentration 0.67 mg/mL
Conjugate HRP
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TYMS
Gene Accession No. P04818
Gene Alias HsT422, Thymidylate synthase, thymidylate synthetase, TMS, TS, TSase, TYMS
Gene Symbols TYMS
Host Species Mouse
Immunogen TYMS Fusion Protein Ag7027
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7298
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.