Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Type II Collagenase from Clostridium histolyticum

Catalog No. MP210050205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210050205 5 g
MP21005023 25 mg
MP21005021 100 mg
MP21005025 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP210050205 Supplier MP Biomedicals™ Supplier No. 0210050205
Add to Cart
Add to Cart

Used for heart, bone, muscle, thyroid, cartilage, and liver cells

  • Source: Clostridium histolyticum
TRUSTED_SUSTAINABILITY

Specifications

Form Brown, lyophilized powder
Product Type Collagenase
Quantity 5 g
Source Clostridium histolyticum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.