Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Typenex™ Typesafe™ 2.0 Segment Piercing Device

Click to view available options
Quantity:
1000/Cs.
500 Units/Box
Description
- Dividers inside device guide blood segment to needle, allowing for easier dispensing of blood
- Eliminates need for scissors, reducing risk of cross contamination and mess in lab
- Sits within test tube, allowing increased steadiness when inserting blood segment
- Segment and device may be disposed upon completing typing and screening
Specifications
Specifications
Color | Green |
For Use With (Application) | Pre-transfusion procedures |
Includes | Dividers |
Product Type | Segment Piercing Device |
Disposable | Single-use |
Quantity | 500 Units/Box |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction