Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Typenex™ Typesafe™ 2.0 Segment Piercing Device
SDP

Catalog No. p-8015280
Click to view available options
Quantity:
1000/Cs.
500 Units/Box
This item is not returnable. View return policy
This item is not returnable. View return policy

Allows to sample a blood segment without using scissors.

  • Dividers inside device guide blood segment to needle, allowing for easier dispensing of blood
  • Eliminates need for scissors, reducing risk of cross contamination and mess in lab
  • Sits within test tube, allowing increased steadiness when inserting blood segment
  • Segment and device may be disposed upon completing typing and screening

Specifications

Color Green
For Use With (Application) Pre-transfusion procedures
Includes Dividers
Product Type Segment Piercing Device
Disposable Single-use
Quantity 500 Units/Box
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.