Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYR Mouse anti-Human, Clone: OCA1/812, Abnova™

Catalog No. 89034540 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-540 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-540 Supplier Abnova Corporation Supplier No. MAB14457
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant human TYR.

  • High specificity
  • High reactivity

Specifications

Antigen TYR
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone OCA1/812
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human TYR.
Dilution Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA and 0.05% azide).
Gene Accession No. P14679
Gene Alias OCA1A/OCAIA/SHEP3
Gene Symbols TYR
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human TYR.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7299
Test Specificity Recognizes a cluster of proteins between 70-80kDa, identified as TYR. Occasionally a minor band at 55kDa is also detected. This antibody shows no cross-reaction with MAGE-1 and tyrosinase-related protein 1, TRP-1/gp75.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.