Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyramine, 98+%

Catalog No. AAJ6099006
Click to view available options
Quantity:
1 g
5 g
25 g

Reagent for fluorescence enzyme immunoassay | CAS: 51-67-2 | C8H11NO | 137.182 g/mol

Tyramine acts as a adrenergic and It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. It Is a vaso active aminoacid found in foods and it It precipitates headaches in some sufferers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tyramine acts as a adrenergic and It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. It Is a vaso active aminoacid found in foods and it It precipitates headaches in some sufferers.

Solubility
Soluble in water (10 mg/ml at 15°C), methanol, DMSO, boiling alcohol (100 mg/ml), and ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-67-2
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00008193
InChI Key DZGWFCGJZKJUFP-UHFFFAOYSA-N
Synonym tyramine, 4-2-aminoethyl phenol, 4-hydroxyphenethylamine, p-tyramine, 2-4-hydroxyphenyl ethylamine, uteramine, tyramin, tyrosamine, tocosine, 4-hydroxyphenylethylamine
PubChem CID 5610
ChEBI CHEBI:15760
IUPAC Name 4-(2-aminoethyl)phenol
SMILES C1=CC(=CC=C1CCN)O

Specifications

Melting Point 160°C to 162°C
Color White to Yellow
Boiling Point 175°C to 181°C (8 mmHg) Hg
Quantity 5 g
Beilstein 1099914
Merck Index 14,9835
Solubility Information Soluble in water (10mg/ml at 15°C),methanol,DMSO,boiling alcohol (100mg/ml),and ethanol.
Formula Weight 137.18
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Tyramine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 200-115-8
RTECSNumber SJ5950000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.