Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYRO3 Mouse anti-Human, Clone: 64CT15.6.2.3.1, Abnova™
Mouse monoclonal antibody raised against human TYRO3.
Supplier: Abnova Corporation MAB12328
Specifications
TYRO3 | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) Immunofluorescence (1:10-1:50) The optimal working dilution should be determined by the end user. | |
BYK/Brt/Dtk/FLJ16467/RSE/Sky/Tif | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
7301 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Immunofluorescence, Immunohistochemistry (PFA fixed) | |
64CT15.6.2.3.1 | |
Mouse monoclonal antibody raised against human TYRO3. | |
In PBS (0.09% sodium azide) | |
TYRO3 | |
Recombinant protein corresponding to human TYRO3. | |
400 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction