Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYRO3 Mouse anti-Human, Clone: OTI4C8, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA600226

Description
TYRO3 (tyrosine-protein kinase receptor TYRO3) is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding to several ligands including TULP1 or GAS6. It regulates many physiological processes including cell survival, migration, and differentiation. Ligand binding at the cell surface induces dimerization and autophosphorylation of TYRO3 on its intracellular domain that provides docking sites for downstream signaling molecules. Following activation by a ligand, TYRO3 plays a role in various processes such as neuron protection from excitotoxic injury, platelet aggregation, and cytoskeleton reorganization.Specifications
TYRO3 | |
OTI4C8 | |
Unconjugated | |
TYRO3 | |
BYK, Dtk, Etk-2, RSE, Rek, Sky, Tif | |
Mouse | |
Affinity Chromatography | |
RUO | |
7301 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Monoclonal | |
0.5 mg/mL | |
PBS with <5% trehalose and 0.05% sodium azide | |
Q06418 | |
TYRO3 | |
Full length human recombit protein of human TYRO3 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction