Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrosinase Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS800P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
12 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS800P1 500 μL Unconjugated Concentrated 200μg/mL
MS800P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS800PABX 200 μL Unconjugated Concentrated 1mg/mL
MS800P 1 mL Unconjugated Concentrated 200μg/mL
MS800RQ 12 mL Unconjugated Ready-to-Use Prediluted
MS800B0 100 μL Biotin Concentrated 200μg/mL
MS800B1 500 μL Biotin Concentrated 200μg/mL
MS800B 1 mL Biotin Concentrated 200μg/mL
MS800P0 100 μL Unconjugated Concentrated 200μg/mL
MS800R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS800P1 Supplier Epredia Supplier No. MS800P1
Add to Cart
Add to Cart

Provide accurate, reproducible results with the Epredia™ Tyrosinase Ab-1, Mouse Monoclonal Antibody.

Tyrosinase is a copper-containing metalloglycoprotein that catalyzes several steps in the melanin pigment biosynthetic pathway; the hydroxylation of tyrosine to L-3,4-dihydroxy-phenylalanine (dopa), and the subsequent oxidation of dopa to dopaquinone. Mutations of the tyrosinase gene occur in various forms of albinism. Tyrosinase is one of the targets for cytotoxic T-cell recognition in melanoma patients.

Host Species: Mouse

Clone: T311

Isotype: IgG2a

Species Reactivity: Human, Dog and Cat. Weakly cross-reacts with Mouse. Others not known.

Epitope: aa 1-433

Immunogen: Recombinant tyrosinase protein

Molecular Weight: 70-80kDa

Positive Control: Melanoma cell lines and Melanoma

Cellular Localization: Cytoplasmic Ab-1 shows no cross-reaction with MAGE-1 and tyrosinase-related protein 1, TRP-1/gp75. Staining of melanomas with Ab-1 showed tyrosinase in melanotic as well as amelanotic variants. Ab-1 is a useful marker for melanocytes and melanomas. Occasionally a minor band at 55kDa is also detected in Western Blotting

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunocytology

Specifications

Antigen Tyrosinase Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone T311
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant tyrosinase protein
Quantity 500 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Canine, Feline, Human
Form Concentrated
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.