Learn More
Description
Host Species: Mouse
Clone: T311
Isotype: IgG2a
Species Reactivity: Human, Dog and Cat. Weakly cross-reacts with Mouse. Others not known.
Epitope: aa 1-433
Immunogen: Recombinant tyrosinase protein
Molecular Weight: 70-80kDa
Positive Control: Melanoma cell lines and Melanoma
Cellular Localization: Cytoplasmic Ab-1 shows no cross-reaction with MAGE-1 and tyrosinase-related protein 1, TRP-1/gp75. Staining of melanomas with Ab-1 showed tyrosinase in melanotic as well as amelanotic variants. Ab-1 is a useful marker for melanocytes and melanomas. Occasionally a minor band at 55kDa is also detected in Western Blotting
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunocytology
Specifications
Specifications
| Antigen | Tyrosinase Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | T311 |
| Concentration | Prediluted |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant tyrosinase protein |
| Quantity | 12 mL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.