Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tyrosinase Antibody (OCA1/812), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311469F
Description
Tyrosinase Monoclonal antibody specifically detects Tyrosinase in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Tyrosinase | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OCA1/812 | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
CMM8, EC 1.14.18.1, LB24-AB, Monophenol monooxygenase, OCA1A, OCAIA, SHEP3, SK29-AB, Tumor rejection antigen AB, tyrosinase, tyrosinase (oculocutaneous albinism IA) | |
Recombinant human Tyrosinase protein (Uniprot: P14679) | |
0.1 mL | |
Lipid and Metabolism | |
7299 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction