Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrosinase Monoclonal Antibody (T311), Abbomax

Catalog No. 502204443
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4443 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4443 Supplier Abbomax Inc Supplier No. 606390
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Melanoma Cellular location: Membrane.

Tyrosinase Monoclonal antibody specifically detects Tyrosinase in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Tyrosinase
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone T311
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene TYR
Gene Accession No. P14679
Gene Alias albino; albino locus protein; ATN; C; CMM8; LB24-AB; monophenol monooxygenase; Oca1; OCA1A; OCAIA; oculocutaneous albinism IA; SHEP3; SK29-AB; skc35; Tumor rejection antigen AB; TYR; Tyrosinase; tyrosinase (oculocutaneous albinism IA); tyrosinase precursor
Host Species Mouse
Immunogen A recombinant protein of human tyrosinase
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7299
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.