Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tyrosinase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589322
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589322 Supplier Invitrogen™ Supplier No. PA589322
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FPRACVSSKN LMEKECCPPW SGDRSPCGQL SGRGSCQNIL LSNAPLGPQF PFTGVDDRES WPSVFYNRTC QCSGNFMGFN CGNCKFGFWG PNCTERRLLV RRNIFDLSAP EKDKFFAYLT LAKHTISSDY VIPIGTYGQM KNGSTPMFND INIYDLFVWM HYYVSMDALL GGSEIWRDID FAHEAPAFLP WHRLFLLRWE QEIQKLTGDE NFTIPYWDWR DAEKCDICTD EYMGGQHPTN PNLLSPASFF SSWQIVCSRL EEYNSHQSLC NGTPEGPLRR NPGNHDKSRT PRLPSSADVE FCLSLTQYES GSMDKAANFS F; Positive Samples: Rat skin; Cellular Location: Melanosome membrane, Single-pass type I membrane protein.

The enzyme encoded by this gene catalyzes the first 2 steps, and at least 1 subsequent step, in the conversion of tyrosine to melanin. The enzyme has both tyrosine hydroxylase and dopa oxidase catalytic activities, and requires copper for function. Mutations in this gene result in oculocutaneous albinism, and nonpathologic polymorphisms result in skin pigmentation variation. The human genome contains a pseudogene similar to the 3' half of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tyrosinase
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TYR
Gene Accession No. P14679
Gene Alias albino; albino locus protein; ATN; C; CMM8; LB24-AB; monophenol monooxygenase; Oca1; OCA1A; OCAIA; oculocutaneous albinism IA; SHEP3; SK29-AB; skc35; Tumor rejection antigen AB; TYR; Tyrosinase; tyrosinase (oculocutaneous albinism IA); tyrosinase precursor
Gene Symbols TYR
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-340 of human Tyrosinase (NP_0003631).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308800, 7299
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.