Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tyrosine Hydroxylase Antibody, Novus Biologicals™

Sheep Polyclonal Antibody has been used in 77 publications
Supplier: Novus Biologicals NB300110SS
Description
Tyrosine Hydroxylase Polyclonal specifically detects Tyrosine Hydroxylase in Human, Mouse, Rat, Amphibian, Avian, Mammal, Rabbit, Reptile samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, Single Cell Western.Specifications
Tyrosine Hydroxylase | |
Polyclonal | |
Unconjugated | |
10 mM HEPES (pH 7.5), 0.15 M NaCl, 0.1 mg/mL BSA, 50% Glycerol with No Preservative | |
DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase | |
Sheep | |
60 kDa | |
0.025 mL | |
Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Prostate Cancer | |
7054 | |
Human, Mouse, Rat, Amphibian, Avian, Mammalia, Rabbit, Reptile | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
Western Blot 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:1000, Immunohistochemistry Free-Floating, Single Cell Western | |
P04177 | |
TH | |
SDS-denatured, native rat tyrosine hydroxylase purified from pheochromocytoma. | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼60 kDa tyrosine hydroxylase protein. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction