Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tyrosine Hydroxylase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$332.50 - $545.00
Specifications
Antigen | Tyrosine Hydroxylase |
---|---|
Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL |
Applications | Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Tyrosine Hydroxylase Polyclonal antibody specifically detects Tyrosine Hydroxylase in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Tyrosine Hydroxylase | |
Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Prostate Cancer, Tyrosine Kinases | |
PBS, pH 7.2, 40% glycerol | |
7054 | |
IgG | |
Affinity purified |
Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
Polyclonal | |
Purified | |
RUO | |
Human | |
DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase | |
This antibody was developed against Recombinant Protein corresponding to amino acids: EFGLCKQNGEVKAYGAGLLSSYGELLHCLSEEPEIRAFDPEAAAVQPYQDQTYQSVYFVSESFSDAKDKLRSYASRIQRPFSVKFDPYTLAIDVLDSPQAVRRSLEGVQDEL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title