Learn More
MilliporeSigma™ Tyrosine Hydroxylase, Mouse, Unlabeled, Clone: 10F8.2,
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Tyrosine Hydroxylase |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 10F8.2 |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal IgG2bλ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide. |
| Gene | Th |
| Gene Accession No. | P04177 |
| Host Species | Mouse |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.