Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tyrosine Hydroxylase, p Ser40 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NB300173
Description
Tyrosine Hydroxylase Polyclonal specifically detects Tyrosine Hydroxylase in Mouse, Rat, Mammal samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Tyrosine Hydroxylase (p Ser40) | |
Polyclonal | |
Unconjugated | |
10 mM HEPES (pH 7.5), 0.15 M NaCl, 0.1 mg/mL BSA, 50% Glycerol with No Preservative | |
DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase | |
Rabbit | |
60 kDa | |
0.1 mL | |
Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Prostate Cancer | |
7054 | |
Mouse, Rat, Mammalia, Other | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
0.1 mg/mL | |
Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Frozen 1:1000 | |
P04177 | |
TH | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Tyrosine Hydroxylase conjugated to KLH. Accession # P04177 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼60 kDa tyrosine hydroxylase protein phosphorylated at Ser40. Some higher molecular weight bands may be detected by the antibody depending upon the brain region being studied, protein loads and the detection methods used. The antibody has three orders of magnitude selectivity over dephospho TH. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction