Learn More
Invitrogen™ Tyrosine Hydroxylase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585356
Description
Keep as concentrated solution. Predicted reactivity: Mouse (92%), Rat (93%), Zebrafish (82%), Japanese Medaka (83%), Xenopus laevis (83%), Dog (96%), Chicken (88%), Bovine (93%). Positive Control: NT2D1, PC-3, U87-MG, SK-N-SH, mouse brain, PC-12, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Tyrosine hydroxylase (TH) is an enzyme involved in the synthesis of catecholamine neurotransmitters dopamine, epinephrine, and norepinephrine. In all species, catecholamine synthesis is regulated by the interaction of TH with a cofactor, tetrahydrobiopterin (BH4). BH4 binds to the TH catalytic domain, resulting in enzymatic activity. Unlike TH in non-primate species, four human TH mRNA splice variants (hTH1-hTH4) have been isolated. These variants are identical in their catalytic domain, but differ in their N-terminal, regulatory domains. TH is also responsible for the conversion of L-tyrosine to L-dopa. TH plays a key role in the physiology of adrenergic neurons. The role of TH in the synthesis of catecholamine neurotransmitters suggests a correlation between the enzyme and a number of neuropathogenic diseases including: Parkinson's disease, schizophrenia, Segawa syndrome, and dystonia, as well as a variety of cardiovascular diseases.
Specifications
Tyrosine Hydroxylase | |
Polyclonal | |
Unconjugated | |
TH | |
dystonia 14; DYT14; DYT5b; EC 1.14.16.2; HGNC:11782; Th; TH isoform 3; TH isoform a; th1; TH-4; The; TY3H; TYH; TYH antibody; Tyrosine 3-hydroxylase; tyrosine 3-monooxygenase; tyrosine hydroxylase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21823, 25085, 280707, 7054 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.21 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P04177, P07101, P17289, P24529 | |
TH | |
Recombinant protein encompassing a sequence within the center region of human Tyrosine Hydroxylase. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Bovine, Rodent | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.