Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrosine, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18607625
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

T1153, 60-18-4, C9H11NO3

Spectrum™ Chemical Tyrosine, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 60-18-4
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00002606
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
IUPAC Name (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.004
Loss on Drying 0.003
Packaging Poly Bottle
Specific Rotation -9.8° to -11.2°
Quantity 100 g
Percent Purity 98.5 to 101.5%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.