Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYRP1 Antibody (SPM611), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311452APC
Description
TYRP1 Monoclonal antibody specifically detects TYRP1 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
TYRP1 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
SPM611 | |
Flow Cytometry | |
b-PROTEIN, CAS25,6-dihydroxyindole-2-carboxylic acid oxidase, Catalase B, CATB, EC 1.14.18, EC 1.14.18.-, EC 1.14.18.1, Glycoprotein 75, GP75DHICA oxidase, Melanoma antigen gp75, TRP-1, TRPTYRP, tyrosinase-related protein 1TRP1, TYRRP | |
Recombinant full-length human TYRP1 protein (Uniprot: P17643) | |
0.1 mL | |
Cancer | |
7306 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction