Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYRP1 Antibody (TYRP1/1564R), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP254446AF750
Description
TYRP1 Monoclonal specifically detects TYRP1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TYRP1 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
TYRP1 | |
Recombinant human TYRP1 protein (Uniprot: P17643) | |
Cancer | |
7306 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
TYRP1/1564R | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
b-PROTEIN, CAS25,6-dihydroxyindole-2-carboxylic acid oxidase, Catalase B, CATB, EC 1.14.18, EC 1.14.18.-, EC 1.14.18.1, Glycoprotein 75, GP75DHICA oxidase, Melanoma antigen gp75, TRP-1, TRPTYRP, tyrosinase-related protein 1TRP1, TYRRP | |
Rabbit | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction