Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYRP1 Antibody (TYRP1/1986) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261600200UG
Description
TYRP1 Monoclonal antibody specifically detects TYRP1 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, SDS-Page.Specifications
TYRP1 | |
Monoclonal | |
1 mg/mL | |
ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, SDS-Page | |
b-PROTEIN, CAS25,6-dihydroxyindole-2-carboxylic acid oxidase, Catalase B, CATB, EC 1.14.18, EC 1.14.18.-, EC 1.14.18.1, Glycoprotein 75, GP75DHICA oxidase, Melanoma antigen gp75, TRP-1, TRPTYRP, tyrosinase-related protein 1TRP1, TYRRP | |
Mouse | |
75 kDa | |
200 μg | |
7306 | |
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
ELISA, Immunohistochemistry (Paraffin), SDS-Page | |
TYRP1/1986 | |
Unconjugated | |
10mM PBS - BSA free with No Preservative | |
TYRP1 | |
Recombinant fragment of human TYRP1 protein (around aa 257-377) (exact sequence is proprietary) | |
Protein A or G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction