Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TYRP1 Monoclonal Antibody (Ta99)

Catalog No. PIMA512293
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512293 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512293 Supplier Invitrogen™ Supplier No. MA512293

Mouse Monoclonal Antibody

MA5-12293 targets Tyrosinase Related Protein 75 in IHC (F) and WB applications and shows reactivity with Human samples. The MA5-12293 immunogen is sK-MEL-23 Melanoma cell line.

This gene encodes a melanosomal enzyme that belongs to the tyrosinase family and plays an important role in the melanin biosynthetic pathway. Defects in this gene are the cause of rufous oculocutaneous albinism and oculocutaneous albinism type III.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TYRP1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone Ta99
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene TYRP1
Gene Accession No. P17643
Gene Alias 5,6-dihydroxyindole-2-carboxylic acid oxidase; B; b-PROTEIN; brown; Brown locus protein; CAS2; Catalase B; CATB; DHICA oxidase; glycoprotein 75; GP75; iris stromal atrophy; isa; melanogenic enzyme; melanoma antigen gp75; MGC120175; MGC149362; OCA3; RP11-3L8.1; TRP; TRP1; TRP-1; truncated Tyrp1; TRY1; TRY4; TRYP1; tyrosinase related protein 1; tyrosinase-related protein 1; tyrosinase-related protein 1 precursor; tyrosine phosphatase 1, same as B (Brown); TYRP; Tyrp1; Tyrp-1; TYRRP
Gene Symbols TYRP1
Host Species Mouse
Immunogen SK-MEL-23 Melanoma cell line.
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7306
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.