Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYRP1, Mouse anti-Human, Clone: TYRP1/3280, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | TYRP1 |
---|---|
Clone | TYRP1/3280 |
Concentration | 0.2 mg/mL |
Dilution | ELISA 2-4 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
Description
TYRP1 Monoclonal specifically detects TYRP1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
TYRP1 | |
0.2 mg/mL | |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
P17643 | |
7306 | |
A recombinant fragment (around aa 257-377) of human TYRP1 protein (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
TYRP1/3280 | |
ELISA 2-4 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
b-PROTEIN, CAS25,6-dihydroxyindole-2-carboxylic acid oxidase, Catalase B, CATB, EC 1.14.18, EC 1.14.18.-, EC 1.14.18.1, Glycoprotein 75, GP75DHICA oxidase, Melanoma antigen gp75, TRP-1, TRPTYRP, tyrosinase-related protein 1TRP1, TYRRP | |
TYRP1 | |
IgG2b κ | |
Protein A or G purified | |
This antibody reacts with It reacts with a 75kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TYRP1). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title