Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TYRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569298
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569298 Supplier Invitrogen™ Supplier No. PA569298
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 100%.

This gene encodes a melanosomal enzyme that belongs to the tyrosinase family and plays an important role in the melanin biosynthetic pathway. Defects in this gene are the cause of rufous oculocutaneous albinism and oculocutaneous albinism type III.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TYRP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TYRP1
Gene Accession No. 0
Gene Alias 5,6-dihydroxyindole-2-carboxylic acid oxidase; B; b-PROTEIN; brown; Brown locus protein; CAS2; Catalase B; CATB; DHICA oxidase; glycoprotein 75; GP75; iris stromal atrophy; isa; melanogenic enzyme; melanoma antigen gp75; MGC120175; MGC149362; OCA3; RP11-3L8.1; TRP; TRP1; TRP-1; truncated Tyrp1; TRY1; TRY4; TRYP1; tyrosinase related protein 1; tyrosinase-related protein 1; tyrosinase-related protein 1 precursor; tyrosine phosphatase 1, same as B (Brown); TYRP; Tyrp1; Tyrp-1; TYRRP
Gene Symbols TYRP1
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of rat TYRP1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298182
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.