Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYRP1 Rabbit anti-Human, Mouse, Unconjugated, Clone: TYRP1/1564R, Abnova™

Catalog No. 89339741 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-741 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-741 Supplier Abnova Corporation Supplier No. RAB00702
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

TYRP1 Monoclonal antibody specifically detects TYRP1 in Human, Mouse samples. It is validated for IHC-P.

Specifications

Antigen TYRP1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TYRP1/1564R
Conjugate Unconjugated
Description tyrosinase-related protein 1
Gene TYRP1
Gene Alias CAS2, CATB, GP75, TRP, TYRP, b-PROTEIN
Host Species Rabbit
Immunogen Recombinant human TYRP1 protein
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7306
Target Species Human, Mouse
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.